Learn More
Mouse anti-Human IgG (H+L) Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Mouse Polyclonal Antibody
Brand:  Novus Biologicals NBP1-72683
369.75 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18283394
  • 511.00 € / 1mg
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
Western Blot 1:500 - 1:2500, ELISA 1:2000 - 1:10000, Immunohistochemistry 1:200-1:1000
Human IgG whole molecule
1 mg
Adaptive Immunity, Immunology
This product was prepared from monospecific polyclonal ascites by immunoaffinity chromatography using Human IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Mouse Serum, Human IgG and Human Serum.
Store at 4C. Do not freeze.
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alkaline Phosphatase (AP)
Mouse
Affinity Purified
RUO
Secondary