missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG1 Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen MH1013
188.10 EUR valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Additional Details : Weight : 0.02000kg
Description
Human IgG1 Monoclonal antibody specifically detects Human IgG1 in Human samples. It is validated for Immunocytochemistry.Specifications
Human IgG1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
IgG gamma 1 | |
500 μL | |
Secondary | |
-20°C | |
IgG1 κ |
Immunocytochemistry | |
HP6069 | |
Unconjugated | |
Mouse | |
purified | |
RUO | |
Human | |
Liquid |