missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Human IgG2, Clone: G18-21, Secondary Antibody, BD

Product Code. 15868498
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15868498 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15868498 Supplier BD Biosciences Supplier No. 555873

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The G18-21 antibody reacts specifically with human IgG2. It does not react with other Ig isotypes.

Host Species: Mouse
Clone: G18-21
Isotype: IgG1, κ
Species Reactivity: Human

ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Human IgG2
Classification Monoclonal
Clone G18-21
Concentration 0.5mg/mL
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Human
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.