missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG2 Fc Secondary Antibody (52G1), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-7082APC
This item is not returnable.
View return policy
Description
IgG2 Fc Monoclonal antibody specifically detects IgG2 Fc in Human samples. It is validated for Flow Cytometry
Specifications
| IgG2 Fc | |
| Monoclonal | |
| APC | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Flow Cytometry | |
| 52G1 | |
| PBS | |
| Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunised with human IgG2 from myeloma patient serum. | |
| 0.2 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction