missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Human IgG3 (Hinge) Secondary Antibody, HRP

Product Code. 17136078
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17136078 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17136078 Supplier Invitrogen™ Supplier No. 053620
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Secondary Antibody

Mouse anti-Human IgG3 (Hinge) Secondary Antibody for ELISA

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG3 (Hinge)
Applications ELISA
Classification Monoclonal
Clone HP6047
Concentration 1 mg/mL
Conjugate HRP
Formulation PBS with 4mg/mL BSA, 40% glycerol and 0.1% Kathon; pH 7.4
Host Species Mouse
Immunogen Human IgG3.
Purification Method purified
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.