missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Anti-Human IgM, Clone: G20-127, PE, Secondary Antibody, BD
Brand: BD Biosciences 551079
This item is not returnable.
View return policy
Description
The G20-127 antibody reacts specifically with the heavy chain of human IgM. It does not react with other heavy chain isotypes.
Host Species: Mouse
Clone: G20-127
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Specifications
| Mouse Anti-Human IgM | |
| G20-127 | |
| Mouse | |
| RUO | |
| Secondary | |
| IgG1 κ |
| Monoclonal | |
| PE-Cyanine5 | |
| 100 Tests | |
| Immunology | |
| Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction