missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgM Fc Secondary Antibody, APC, Invitrogen™
Mouse Monoclonal Secondary Antibody
Brand: Invitrogen SA119468
Code : LD
Additional Details : Weight : 0.01000kg
Description
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.Specifications
Human IgM Fc | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.2% BSA and 15mM sodium azide; pH 7.4 | |
Purified human IgM. | |
100 μg | |
Secondary | |
4° C, store in dark, DO NOT FREEZE! | |
IgG1 |
Flow Cytometry | |
CH2 | |
APC | |
Mouse | |
Size-exclusion chromatography | |
RUO | |
Human | |
Liquid |