missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Mouse IgG1[a], Clone: 10.9, Secondary Antibody, BD
Description
This antibody is specific for mouse IgG1 of the Igh-C[a] and the c, d, e, f, g, h, n, o, and p haplotypes (e.g. BALB/c, C58, A, AKR, CBA, C3H, DBA/1, DBA/2, NZB, SWR). It has been reported not to react with IgG1 of the Igh-C[b] haplotype (e.g. C57BL/6, C5
Host Species: Mouse
Clone: 10.9
Isotype: IgG2a, κ
Species Reactivity: Mouse
ELISA, Flow Cytometry
Specifications
Specifications
| Antigen | Mouse anti-Mouse IgG1[a] |
| Classification | Monoclonal |
| Clone | 10.9 |
| Concentration | 0.5mg/mL |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 0.5 mg |
| Regulatory Status | RUO |
| Research Discipline | Immunology |
| Primary or Secondary | Secondary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?