missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Mouse IgG2a[a], Clone: 8.3, Secondary Antibody, BD

Product Code. 15827248
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15827248 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15827248 Supplier BD Biosciences Supplier No. 553501

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The 8.3 antibody reacts specifically with mouse IgG2a of Igh-Ca and the d, e, f, g, h, j, n, and o haplotypes (eg, BALB/c, C58, and A, AKR, CBA, C3H/He, NZB). It does not react with IgG2a of Igh-Cb nor the c and p haplotypes (eg, C57BL/6, C57BL/10, SJL, D

Host Species: Mouse
Clone: 8.3
Isotype: IgG2a, κ
Species Reactivity: Mouse

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Mouse IgG2a[a]
Classification Monoclonal
Clone 8.3
Concentration 0.5mg/mL
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Mouse
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.