missing translation for 'onlineSavingsMsg'
Learn More

Mouse Anti-Rabbit IgM, Clone: 367.2, Secondary Antibody, BD

Product Code. 15826438
Change view
Click to view available options
Quantity:
0.25 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15826438 0.25 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15826438 Supplier BD Biosciences Supplier No. 550938

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The 367.2 antibody reacts specifically with rabbit IgM. It does not react with rabbit IgG or IgA isotypes.

Host Species: Mouse
Clone: 367.2
Isotype: IgG1 κ
Species Reactivity: Rabbit
Immunogen: Purified rabbit IgM

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse Anti-Rabbit IgM
Classification Monoclonal
Clone 367.2
Concentration 0.5mg/mL
Conjugate Biotin
Host Species Mouse
Quantity 0.25 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Rabbit
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.