Learn More
Invitrogen™ Mouse anti-Rat IgD Secondary Antibody, Biotin
Mouse Monoclonal Secondary Antibody
Brand:  Invitrogen™ MA516809
Product Code. 15296049
  • 760.00 € / 500µg
Description
Mouse anti-Rat IgD Secondary Antibody for ELISA
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rat IgD
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide
IR731 myeloma protein.
500 μg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA
MARD-3
Biotin
Mouse
Affinity chromatography
RUO
Rat
Secondary Antibody
IgG1