missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse anti-Rat IgG1 Secondary Antibody, NovaFluor™ Blue 760, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
181.00 € - 399.00 €
Specifications
| Antigen | CD45 |
|---|---|
| Clone | 2D1 |
| Concentration | 0.8 μg/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
30055983
|
Invitrogen™
H005T03B11-A |
100 Tests |
399.00 €
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30054910
|
Invitrogen™
H005T02B11-A |
25 Tests |
181.00 €
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Rat IgG1 Monoclonal antibody specifically detects Rat IgG1 in Human samples. It is validated for Flow CytometrySpécification
| CD45 | |
| 0.8 μg/Test | |
| Monoclonal | |
| Mouse | |
| Human | |
| IgG1 κ | |
| Affinity chromatography |
| 2D1 | |
| Flow Cytometry | |
| NovaFluor Blue 760 | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title