missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Rat IgG2a, Clone: RG7/1.30, Secondary Antibody, BD

Product Code. 15847458
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15847458 0.5 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15847458 Supplier BD Biosciences Supplier No. 553893

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Rg7/1.30 antibody reacts specifically with the Fc region of rat IgG2a. It does not react with other Ig isotypes.

Host Species: Mouse
Clone: RG7/1.30
Isotype: IgG2b, κ
Species Reactivity: Rat
Immunogen: Pooled rat IgG

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse anti-Rat IgG2a
Classification Monoclonal
Clone RG7/1.30
Concentration 0.5mg/mL
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Secondary
Target Species Rat
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.