missing translation for 'onlineSavingsMsg'
Learn More

Mouse anti-Rat IgG2a, Clone: RG7/1.30, biotin, Secondary Antibody, BD

Brand:  BD Biosciences 553894

Product Code. 15857458

  • ONLINE EXCLUSIVE PRICE
    347.00 €
    333.40 € / Each
    Save 13.60 €
    3% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The Rg7/1.30 antibody reacts specifically with the Fc region of rat IgG2a. It does not react with other Ig isotypes.

Host Species: Mouse
Clone: RG7/1.30
Isotype: IgG2b, κ
Species Reactivity: Rat
Immunogen: Pooled rat IgG

ELISA, Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse anti-Rat IgG2a
RG7/1.30
Biotin
0.5 mg
Immunology
Rat
Monoclonal
0.5mg/mL
Mouse
RUO
Secondary
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.