missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse anti-Rat Kappa Light Chain Secondary Antibody, PE

Product Code. 15245744
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15245744 100 μg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15245744 Supplier Invitrogen™ Supplier No. A15428
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Secondary Antibody

Mouse anti-Rat Kappa Light Chain Secondary Antibody for ELISA, Flow

Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rat Kappa Light Chain
Applications ELISA, Flow Cytometry
Classification Monoclonal
Clone K4F5
Concentration 0.1 mg/mL
Conjugate PE
Formulation PBS with <0.1% sodium azide; pH 7.5
Gene Alias Ig kappa chain, Igk, Ig-kappa
Host Species Mouse
Immunogen This mouse IgG1-kappa class-switch variant monoclonal antibody reacts with rat kappa light chains and can be used for identification and enumeration of rat kappa light chain+ cells by flow cytometry.
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse, Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.