missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse BMP-7 Recombinant Protein, PeproTech®

Product Code. 30245220
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 30245220

Brand: Gibco™ 315432UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extra skeletal site. Based on its expression early in embryogenesis, BMP7 has a proposed role in early development. In addition, the fact that BMP7 is closely related to BMP5 and BMP6 has lead to speculation of possible bone inductive activity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P23359
For Use With (Application) Functional Assay
Formulation Protein with no preservative
Gene ID (Entrez) 12162
Molecular Weight (g/mol) 31.5 kDa
Name BMP-7
Quantity 2 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias BMP; BMP7; BMP-7; bone moorphogenic protein-7; Bone morphogenetic protein; bone morphogenetic protein 7; bone morphogenetic protein 7 (osteogenic protein 1); bone morphogenetic protein 7 preproprotein; bone morphogenic protein 7; Eptotermin alfa; OP1; OP-1; Osteogenic protein 1; RP11-560A15.5
Gene Symbol Bmp7
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Sequence STGGKQRSQN RSKTPKNQEA LRMASVAENS SSDQRQACKK HELYVSFRDL GWQDWIIAPE GYAAYYCEGE CAFPLNSYMN ATNHAIVQTL VHFINPDTVP KPCCAPTQLN AISVLYFDDS SNVILKKYRN MVVRACGCH
Activity Determined by its ability to induce alkaline phosphatase production by ADTC-5 cells.
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.