missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse CD3 epsilon Heterodimer Recombinant Protein

Product Code. 17946101
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 17946101

Brand: Invitrogen™ RP88216

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (200 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.

CD3E and CD3D Heterodimer Protein is produced by mammalian expression system and are involved in mediating signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P04235, P22646
Concentration 0.25 mg/mL
For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation PBS with 5% mannitol, 5% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Gene ID (Entrez) 12500, 12501
Molecular Weight (g/mol) 77.53 kDa
Name Mouse CD3 epsilon Heterodimer
pH Range 7.4
Purification Method Purified
Quantity 50 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Common Name CD3e/CD3d
Gene Symbol CD3D, CD3E
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence Mouse CD3D, Met1-Ala105 (Accession # P04235) with a C-terminal Flag-tag hIgG1 Fc region and mouse CD3E, Met1-Asp108 (Accession # P22646) with a C-terminal His-tag hIgG Fc region were co-expressed and purified as a mouse CD3D & CD3E heterodimer
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade ≥90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.