missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse CD80 ELISA Kit
Mouse CD80 quantitates mouse CD80 in serum, plasma, supernatant.
644.00 € - 4635.00 €
Specifications
Accession Number | Activation B7-1 antigen; B7; B7 protein; B7.1; B71; B7-1; B7-1 protein; BB1; B-lymphocyte activation antigen B7; Cd28l; CD28LG; CD28LG1; CD80; Cd80 a rat homolog of the human CD28/CTLA - 4 ligand (B7-1); CD80 anitgen; CD80 antigen; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); Cd80 molecule; CD80 protein precursor; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; LAB7; Ly53; Ly-53; MIC17; sCD 80; sCD80; soluble CD 80; soluble CD80; T-cell costimulatory molecule; T-cell co-stimulatory protein B7-1; T-lymphocyte activation antigen CD80; TS/A-1; TSA1 |
---|---|
Assay Range | 28 to 7,000 pg/mL |
Assay Sensitivity | 28 pg/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17185609
|
Invitrogen™
EM16RBX5 |
5 x 96 Tests |
2635.00 €
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
17174863
|
Invitrogen™
EM16RB |
96 Tests |
644.00 €
Pack of 96 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
18527864
|
Invitrogen™
EM16RBX10 |
10 x 96 Tests |
4635.00 €
Pack of 10 |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Mouse CD80 quantitates mouse CD80 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse CD80.
CD80 (B7-1) and CD86 (B7-2) are ligands of T cell critical costimulatory molecule CD28, and of an inhibitory receptor CTLA-4 (CD152). Both B7 molecules are expressed on professional antigen-presenting cells and are essential for T cell activation, and both molecules can also substitute for each other in this process. CD80 is rapidly induced on the surface of in vitro activated B cells, with Epstein Barr Virus (EBV) transformed B cell lines, with Burkitts lymphoma cell lines, with freshly isolated follicular B lymphoma cells, T cells, and monocytes. The B-lymphocyte activation antigen B7-1 provides regulatory signals for T lymphocytes as a consequence of binding to the CD28 and CTLA4 ligands of T cells. Diseases associated with CD80 dysfunction include gallbladder squamous cell carcinoma and myocarditis.Specifications
Activation B7-1 antigen; B7; B7 protein; B7.1; B71; B7-1; B7-1 protein; BB1; B-lymphocyte activation antigen B7; Cd28l; CD28LG; CD28LG1; CD80; Cd80 a rat homolog of the human CD28/CTLA - 4 ligand (B7-1); CD80 anitgen; CD80 antigen; CD80 antigen (CD28 antigen ligand 1, B7-1 antigen); Cd80 molecule; CD80 protein precursor; costimulatory factor CD80; costimulatory molecule variant IgV-CD80; CTLA-4 counter-receptor B7.1; LAB7; Ly53; Ly-53; MIC17; sCD 80; sCD80; soluble CD 80; soluble CD80; T-cell costimulatory molecule; T-cell co-stimulatory protein B7-1; T-lymphocyte activation antigen CD80; TS/A-1; TSA1 | |
28 pg/mL | |
ELISA | |
Q00609 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
CD80 (B7-1) | |
4 hr 45 min |
28 to 7,000 pg/mL | |
Biotin | |
Colorimetric Microplate Reader | |
CD80 | |
<10% | |
HRP | |
2°C to 8°C | |
1 hr 20 min |