missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CEACAM-16 (AAI41088) VersaClone cDNA

Product Code. 18799767
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18799767 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18799767 Supplier R&D Systems™ Supplier No. RDC2180
  • ONLINE EXCLUSIVE PRICE
    341.00 €
    322.35 € / 10µg
    Save 18.65 €
    5% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

R&D Systems™ Mouse CEACAM-16 (AAI41088) VersaClone cDNA contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation.

The Mouse CEACAM-16 (AAI41088) VersaClone cDNA clone contains the complete ORF for CEACAM16, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The CEACAM16 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Specifications

Gene mCeacam16
Accession Number AAI41088
Concentration 10 μg at 0.2 μg/μL
Gene ID (Entrez) 388551
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.