missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CLEC9a (NP_766320) VersaClone cDNA

Product Code. 18716398
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18716398 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18716398 Supplier R&D Systems™ Supplier No. RDC2809
  • ONLINE EXCLUSIVE PRICE
    328.00 €
    309.75 € / 10µg
    Save 18.25 €
    5% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Accession Number NP_766320
Gene ID (Entrez) 283420
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.