missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse Collagen I ELISA Kit

Product Code. 30245254
Click to view available options
:
96 Tests
This item is not returnable. View return policy

Product Code. 30245254

Brand: Invitrogen™ EEL218

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

Collagen I is a Type I collagen with a triple helix structure comprised of two alpha-1 chains and one alpha-2 chain. Collagen I is a member of group I collagen (fibril-forming collagen) found in most connective tissues, and is abundant in bone, cornea, dermis and tendon. Mutations in the COL1A2 gene encoding the alpha-2 chain are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIB, recessive Ehlers-Danlos syndrome Classical type, idiopathic osteoporosis, and atypical Marfan syndrome. Symptoms associated with mutations in the COL1A2 gene which codes for the alpha-2 chain, however, tend to be less severe than mutations in the COL1A1 gene, reflecting the different role of alpha-2 chains in matrix integrity. Reciprocal translocations between chromosomes 17 and 22, where Collagen Type 1 genes and the gene for platelet derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P11087, Q01149
Assay Range 0.31 to 20 ng/mL
Assay Sensitivity 0.19 ng/mL
Conjugate HRP
Detection Method Colorimetric
Immunoassay Kit Format ELISA Microplate (Dismountable)
Product Type ELISA
Sample Type Plasma, Serum
Target Species Mouse
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 12842, 12843
Gene Symbol COL1A1, COL1A2
Interassay CV <10
Intraassay CV <10
Kit Contents Micro ELISA Plate (Dismountable), Reference Standard, Concentrated Biotinylated Detection Ab(100x), Concentrated HRP Conjugate (100x), Reference Standard & Sample Diluent, Biotinylated Detection Ab Diluent, HRP Conjugate Diluent, Concentrated Wash Buffer(25x), Substrate Reagent, Stop Solution, Plate Sealer, Product Description, Certificate of Analysis
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma: 100 μL, Serum: 100 μL
Storage Requirements Refer to product documentation for component specific storage temperature.
Target Collagen I
Test Time 1 hr. 20 min.
Total Assay Time 3 hr. 30 min.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.