missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse CTRP3/C1qTNF3/CORS26 (NP_112150) VersaClone cDNA

Product Code. 18777278
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18777278 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18777278 Supplier R&D Systems™ Supplier No. RDC2139
  • ONLINE EXCLUSIVE PRICE
    341.00 €
    322.35 € / 10µg
    Save 18.65 €
    5% Off
 Restricted Item
Estimated Shipment: 23-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

R&D Systems™ Mouse CTRP3/C1qTNF3/CORS26 (NP_112150) VersaClone cDNA is a member of the C1q family of proteins.

The Mouse CTRP3/C1qTNF3/CORS26 (NP_112150) VersaClone cDNA clone contains the complete ORF for C1qTNF3/CORS26/CTRP3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The C1qTNF3/CORS26/CTRP3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Specifications

Gene mC1qtnf3
Accession Number NP_112150
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 754bp
pH 8.5
Gene ID (Entrez) 114899
Gene Alias C1ATNF3, C1q and tumor necrosis factor related protein 3, C1qTNF3, Cartducin, cartonectin, collagenous repeat-containing sequence of 26-kDa, complement C1q tumor necrosis factor-related protein 3, complement-c1q tumor necrosis factor-related protein 3, Corcs, Cors, Cors-26, CORS26, CTRP32310005P21Rik, FLJ37576, Secretory protein CORS26
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.