missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Mouse CXCL2 ELISA Kit

Product Code. 16831616 Shop All Proteintech Products
Change view
Click to view available options
Target Species:
Mouse
Unit Size:
96 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Target Species unitSize
16831616 Mouse 96 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16831616 Supplier Proteintech™ Supplier No. KE1002296T

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

KE10022 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The CXCL2 ELISA kit is to be used to detect and quantify protein levels of endogenous CXCL2.

  • The assay recognizes mouse CXCL2. An antibody specific for CXCL2 has been pre-coated onto the microwells.
  • The CXCL2 protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody of biotinylated specific for CXCL2 is added to detect the captured CXCL2 protein.
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent.
  • Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450nm with the correction wavelength set at 630 nm
  • Cxcl2, also known as macrophage inflammatory protein 2-alpha (Mip2-alpha), is a member of CXC chemokine family that binds to CXC chemokine receptor 2 (Cxcr2).
  • Cxcl2 is mainly produced by macrophages, endothelial cells, epithelial cells and tumor cells. Cxcl2 plays important roles in various biological progresses such as angiogenesis, inflammation, immune response and cancer biological behaviors.
  • Under inflammatory conditions in CNS, microglia and astrocytes may secrete Cxcl2 as well as other chemokines, which induces chemotaxis and infiltration of circulatory neutrophils.
  • Indeed, previous studies suggest that Cxcl2 is involved in Alzheimer disease, multiple sclerosis and brain ischemia.

Specifications

Accession Number NP_033166
Assay Range 31.25 to 500 pg/mL
Assay Sensitivity 20.5 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Mouse
Gene ID (Entrez) 20310
Host Species Mouse
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID P10889
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human TIMP1 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Sulphuric Acid, Tetramethylbenzidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.