missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Dopamine D3 R/DRD3 (NP_031903.1) VersaClone cDNA

Product Code. 18790377
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18790377 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18790377 Supplier R&D Systems™ Supplier No. RDC3786
  • ONLINE EXCLUSIVE PRICE
    365.00 €
    344.40 € / 10µg
    Save 20.60 €
    5% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Gene Dopamine D3R/DRD3
Accession Number NP_031903.1
Gene ID (Entrez) 1814
Gene Alias D(3) dopamine receptor, D3DR, Dopamine D3 receptor, Dopamine D3R, dopamine receptor D3, DRD3, essential tremor 1, ETM1, FET1, MGC149204, MGC149205
Content And Storage Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Product Line R&D Systems
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.