missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse Granzyme B ELISpot Kit
Specifications
Specifications
| Antigen | Granzyme B |
| Includes | One 96-well plate coated w/monoclonal Ab for mouse Granzyme B, Biotinylated detection Ab, Streptavidin-AP, Buffers, BCIP/NBT chromogen, Positive control |
| Quantity | 1 Kit |
| Sample Type | Whole Cells |
| Synonym | C11, CCPI, CGL1, CGL-1, CSPB, CSP-B, CTLA1, CTLA-1, CTSGL1, Fragmentin-2, granzyme B, Granzyme-2, GranzymeB, GRB, GrzB, GZMB, HLP, SECT |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Gene ID (Entrez) | 3002 |
| Gene Symbol | Gzmb |
| For Use With (Application) | Diagnostics |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?