Learn More
Mouse IgG VisUCyte HRP Polymer Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  R&D Systems VC001-125
 View more versions of this product
Product Code. 18764388
  • 1370.00 € / 125mL
Description
IgG Polyclonal specifically detects IgG in Mouse samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG
Polyclonal
Immunohistochemistry
Immunoglobulin G, ImmunoglobulinG
Mouse IgG
125 mL
Secondary
Detects heavy ( gamma ) chains on mouse IgG and light chains on all mouse immunoglobulins, based on IEP. No cross-reactivity is observed with non-immunoglobulin mouse serum proteins or serum proteins from bovine, horse, human, pig or rabbit, based on IEP. VisUCyteÖ HRP Polymer provides high sensitivity with low background in IHC.
6 months from date of receipt, 2 to 8 degreesC as supplied.
Immunohistochemistry
HRP Polymer
Supplied as a solution in PBS with preservative.
Goat
Affinity Purified
RUO
3500