Learn More
Mouse IgG1 Isotype Control (W3/25), Invitrogen™
Mouse Isotype Control
Brand: Invitrogen MA133304
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is suitable as a negative control for human, bovine, ovine, porcine, equine, canine, lapine and guinea-pig tissues tissues. It is not suitable for use as a negative control on rat tissues.
Mouse IgG1 is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG1 is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG1 is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.Specifications
Mouse IgG1 | |
W3/25 | |
Unconjugated | |
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183 | |
Proprietary immunogen - Contact ABR for details. | |
250 μg | |
Isotype Control (Primary) | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
Flow Cytometry, Immunohistochemistry (Frozen) | |
1 mg/mL | |
PBS with no preservative | |
Mouse | |
Protein A | |
RUO | |
Rat | |
Liquid |