missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IgG2a Isotype Control, PE-Texas Red

Product Code. 10048682
Change view
Click to view available options
Quantity:
500 μL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10048682 500 μL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10048682 Supplier Invitrogen™ Supplier No. MG2A17
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Mouse Monoclonal Isotype Control

Mouse IgG2a Isotype Control for Flow

Mouse IgG2a is a subtype of immunoglobulin G antibody that is produced by B cells in response to an antigen. It is the second most abundant IgG subtype in the serum and plays a crucial role in the adaptive immune response. Mouse IgG2a is composed of two heavy chains and two light chains, and it is involved in opsonization, complement activation, and antibody-dependent cell-mediated cytotoxicity. It is also involved in the regulation of immune responses, including the activation of T cells and the inhibition of B cell activation. Mouse IgG2a is widely used in research and diagnostic applications, including ELISA, Western blotting, and immunohistochemistry. It is also used in therapeutic applications, such as immunotherapy and vaccine development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG2a
Applications Flow Cytometry
Classification Monoclonal
Conjugate PE-Texas Red
Formulation PBS with 0.1% sodium azide
Gene Alias IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Host Species Mouse
Purification Method Purified
Quantity 500 μL
Regulatory Status ASR
Whole Molecule Whole Antibody
Target Species Not Applicable
Content And Storage 2-8°C
Product Type Isotype Control
Form Liquid
Isotype IgG2a
No. of Reactions 50 Tests
Shipping Condition Wet Ice
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.