missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-12/IL-23 p40 (NP_001290173) VersaClone cDNA

Product Code. 18705399
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18705399 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18705399 Supplier R&D Systems™ Supplier No. RDC2453
  • ONLINE EXCLUSIVE PRICE
    336.00 €
    317.10 € / 10µg
    Save 18.90 €
    5% Off
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sequence verified insert equipped with translated sequences for the insert and entire plasmid DNA.

  • The Mouse IL-12/IL-23 p40 (NP_001290173) VersaClone cDNA clone contains the complete ORF for IL-12/IL-23 p40, along with a Kozak consensus sequence for optimal translation initiation
  • The gene insert is flanked with convenient multiple cloning sites, which can be used to easily cut and transfer the gene cassette into your desired expression vector
  • The IL-12/IL-23 p40 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
TRUSTED_SUSTAINABILITY

Specifications

Gene mIL-12/IL-23p40
Accession Number NP_001290173
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Gene ID (Entrez) 3593
Content And Storage Store the unopened product at -20 to -80°C
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.