missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse IL-33 DuoSet ELISA

Product Code. 30021759
Click to view available options
Quantity:
15 96-well Plates
5 96-well Plates
Unit Size:
1 set
Pack of 5
This item is not returnable. View return policy

Product Code. 30021759

Brand: R&D Systems™ DY3626

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse IL-33 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse IL-33 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse IL-33.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 15 96-well Plates
Conjugate Biotin
Assay Range 15.60 to 1000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 77125
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.