Learn More
R&D Systems™ Mouse Integrin alpha 3 beta 1/VLA-3 Recombinant Protein, Carrier-Free
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 9374-A3-050
Includes: For Research Use Only
Description
The Recombinant Mouse Integrin alpha 3 beta 1/VLA-3 Protein is derived from CHO. The Recombinant Mouse Integrin alpha 3 beta 1/VLA-3 Protein has been validated for the following applications: Bioactivity.Order Info
Ships at ambient temperature.
Specifications
| Q62470-1 (Integrin alpha 3) & P09055-1 (Integrin beta 1) | |
| 3675 | |
| 50μg | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
| <0.10 EU per 1μg of the protein by the LAL method. | |
| Recombinant protein | |
| Yes |
| Lyophilized from a 0.2μm filtered solution in PBS | |
| 116 kDa (Integrin alpha 3), 86 kDa (Integrin beta 1) | |
| Chinese Hamster Ovary cell line,CHO-derived mouse Integrin alpha 3 beta 1/VLA-3 protein, (N-terminus) Mouse Integrin alpha 3 (Phe33-Glu993) Accession &Num; Q62470-1 His-Pro GGGSGGGS Acidic Tail 6-His tag Mouse Integrin beta 1 (Gln21-Asp728) Accession &Num; P09055-1 His-Pro GGGSGGGS Basic Tail (C-terminus) | |
| RUO | |
| 150 to 900ng/mL | |
| Unconjugated |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.