missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse Liver Whole Tissue Lysate (Adult Whole Normal)

Product Code. 18705923
Change view
Click to view available options
Quantity:
0.25 mL
Unit Size:
0.25mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18705923 0.25 mL 0.25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18705923 Supplier Novus Biologicals™ Supplier No. NB800PC17
  • ONLINE EXCLUSIVE PRICE
    339.00 €
    320.25 € / 0.25mL
    Save 18.75 €
    5% Off
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The Mouse Liver Whole Tissue Lysate (Adult Whole Normal) is derived from mouse. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Ready-to-Use Lysates Are Especially Prepared as Positive Controls for Separation by SDS-PAGE and Subsequent Western Blot Analysis. Lysates Are Prepared in Denaturing Buffer WITHOUT Dissociating Agents (i.e. no 2-Mercaptoethanol or Dithiothreitol Has Been Added). Heat Lysate to 95 C for 5 Minutes and Rapidly Cool. If Dissociating Conditions Are Desired, Add Reducing Agent Prior to Heating. The Recommended Loading Volume / Lane is 10-20 μL Depending on The Size Format of Your Gel.
Tissue Digestive/Waste
Quantity 0.25 mL
Storage Buffer 1X SDS-PAGE Sample Buffer (62.5 mM Tris HCl, 2% SDS, 10% Glycerol and 0.005% bromophenol blue, pH 6.8), 10% (v/v) Glycerol
Purity Multi-step
Test Specificity Normal Mouse Liver whole cell extract lysate
Cellular Fraction Whole
Tissue Condition Normal
Host Species Mouse
Concentration 1 mg/mL
Product Type Mouse, Liver Whole Tissue Lysate (Adult Normal)
Content And Storage Store at -70°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.