missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse M-CSF Recombinant Protein, PeproTech®

Product Code. 17882003
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17882003 50 μg 50µg
17812423 100 μg 100µg
17872233 10 μg 10µg
17812133 1 mg 1mg
17812113 250 μg 250µg
17822333 2 μg 2µg
17862003 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 17882003 Supplier Gibco™ Supplier No. 3150250UG
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine M-CSF is a 36.4 kDa homodimeric protein consisting of two 156 amino acid polypeptide subunits. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

M-CSF (Macrophage colony-stimulating factor, CSF-1) is a survival factor essential for the proliferation and development of monocytes, macrophages, and osteoclast progenitor cells. M-CSF also induces VEGF (vascular endothelial growth factor) secretion by macrophages, thereby mediating mobilization of endothelial progenitor cells and neovascularization. M-CSF is present as several bioactive isoforms that differ in potency and stability. The full-length protein is synthesized as a membrane-spanning protein that can be expressed on the cell surface or further cleaved and modified in the secretory vesicle. Further, M-CSF is a disulfide-bonded homodimer which is processed into one of two isoforms, a glycoprotein or a proteoglycan that has been modified by the addition of chondroitin sulfate to each subunit. Binding of M-CSF to its receptor, c-Fms (CSF-1R or CD115) induces dimerization of the receptor followed by internalization and degradation of the complex. Functionally, M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P07141
For Use With (Application) Bioactivity, Control, ELISA Standard, Western Blot Control
Formulation Protein with no preservative
Gene ID (Entrez) 12977
Molecular Weight (g/mol) 36.4 kDa
Name Mouse M-CSF
Purification Method Purified
Quantity 50 μg
Source E. coli
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <1 EU/μg
Gene Alias C87615; colony stimulating factor 1; colony stimulating factor 1 (macrophage); colony-stimulating factor-1 splice variant; CSF1; CSF-1; Csfm; H-MCSF; lanimostim; macrophage colony stimulating factor 1; macrophage colony stimulating factor alpha; macrophage colony stimulating factor beta; macrophage colony-stimulating factor; macrophage colony-stimulating factor 1; macrophage colony-stimulating factor beta; macrophage-colony stimulating factor alpha; MCSF; MCSF alpha; MCSFBETA; MGC31930; M-MCSF; op; osteopetrosis; Processed macrophage colony-stimulating factor 1
Common Name M-CSF
Gene Symbol Csf1
Biological Activity The ED50 as determined by the dose-dependent stimulation of the proliferation of M-NFS-60 cells is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MKEVSEHCSH MIGNGHLKVL QQLIDSQMET SCQIAFEFVD QEQLDDPVCY LKKAFFLVQD IIDETMRFKD NTPNANATER LQELSNNLNS CFTKDYEEQN KACVRTFHET PLQLLEKIKN FFNETKNLLE KDWNIFTKNC NNSFAKCSSR DVVTKP
Content And Storage -20°C
Activity The ED50 as determined by the dose-dependent stimulation of the proliferation of M-NFS-60 cells is ≤ 1.0 ng/ml, corresponding to a specific activity of ≥ 1 x 10^6 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.