Learn More
R&D Systems™ Mouse Oct-3/4 (NP_038661.2) VersaClone cDNA
Product Code. 18730517
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
18730517 10 μg 10µg
1 options
Product Code. 18730517 Supplier R&D Systems™ Supplier No. RDC3642
  • ONLINE EXCLUSIVE PRICE
    287.00 €
    270.90 € / 10µg
    Save 16.10 €
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
Specifications
Gene Oct-3/4
Accession Number NP_038661.2
Gene ID (Entrez) 5460
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.