missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse Pancreas Whole Tissue Lysate (Adult Whole Normal)

Product Code. 18788903
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18788903 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18788903 Supplier Novus Biologicals™ Supplier No. NB82059665
  • ONLINE EXCLUSIVE PRICE
    396.00 €
    373.80 € / 1mg
    Save 22.20 €
    5% Off
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The Mouse Pancreas Whole Tissue Lysate (Adult Whole Normal) is derived from mouse. It has been validated for the following applications: Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot
Tissue Digestive/Waste
Quantity 1 mg
Storage Buffer HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Purity Protein
Test Specificity Total Protein - Mouse Normal Tissue: Pancreas
Cellular Fraction Whole
Tissue Condition Normal
Cross Reactivity Mouse
Host Species Mouse
Concentration 5 mg/mL
Product Type Mouse Pancreas Whole Tissue Lysate (Adult Normal)
Content And Storage -80°C. Avoid freeze-thaw cycles.
Life Stage Adult
Protein State Native
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.