missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human Perforin QuicKit ELISA

Product Code. 30306088
Click to view available options
:
1 Kit
This item is not returnable. View return policy

Product Code. 30306088

Brand: R&D Systems™ QK8011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

one step, 80-minute solid phase ELISA designed to measure human Perforin levels in cell culture supernates, serum, plasma, and cell lysates

TRUSTED_SUSTAINABILITY

Specifications

Specificity Natural and recombinant human Perforin.
Target Species Human
Gene ID (Entrez) 5551
Quantity 1 Kit
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFP, PFPcytolysin, PRF1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.