missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse PLA2R1 (NP_032893.1) VersaClone cDNA

Product Code. 18700237
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18700237 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18700237 Supplier R&D Systems™ Supplier No. RDC3097
  • ONLINE EXCLUSIVE PRICE
    1215.00 €
    1147.65 € / 10µg
    Save 67.35 €
    5% Off
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Contains gene inserts that are fully sequenced and produced with high quality

TRUSTED_SUSTAINABILITY

Specifications

Gene PLA2R1
Accession Number NP_032893.1
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 4477 bp
Gene ID (Entrez) 22925
Gene Alias 180 kDa secretory phospholipase A2 receptor, CLEC13C, CLEC13CC-type lectin domain family 13 member C, phospholipase A2 receptor 1, 180 kDa, PLA2G1R, PLA2IR, PLA2-RM-type receptor, PLA2Rphospholipase A2 receptor 1, 180 kD, secretory phospholipase A2 receptor
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.