missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse PPAR delta (aa 1-14) Synthetic Peptide
Description
Murine PPAR delta (residues 1-14) Synthetic Peptide, PEP-026, from Invitrogen.
PEP-026 is a 14 amino acid (amino acids 1-14) synthetic peptide whose sequence is derived from murine PPAR delta and is completely unique to PPAR delta. A COOH-terminal cysteine residue was added for conjugation to KLH. The sequence of this peptide is (amino to carboxy terminus): M1-E-Q-P-Q-E-E-T-P-E-A-R-E-E14-CThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and PPAR delta, catalog # PA1-823A. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody.Reconstitute with 0.1 ml of distilled water.
Specifications
Specifications
| Gene ID (Entrez) | 19015 |
| Species | Mouse |
| Purification Method | purified |
| Accession Number | P35396 |
| Common Name | Mouse PPAR delta (aa 1-14) |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | peptide with no preservative |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?