missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse/Rat Caveolin 3 (aa 1-18) Synthetic Peptide

Produktkod. 11504740
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
50 μg
Förpackningsstorlek
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
11504740 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11504740 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' PEP081

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Synthetic Peptide

Mouse and Rat Caveolin-3 (residues 1-18) Synthetic Peptide, PEP-081, from Invitrogen.

PEP-081 is a 19 amino acid synthetic peptide whose sequence corresponds to residues 1-18 of mouse and rat caveolin-3 with a carboxy terminal cysteine added for carrier protein conjugation. This sequence is ∼85% conserved in human CAV3. The sequence of this peptide is (amino to carboxy terminus: M (1) M T E E H T D L E A R I I K D I H (18) CThis peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and endogenous caveolin-3, catalog # PA1-066. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.
TRUSTED_SUSTAINABILITY

Specifikationer

Gene ID (Entrez) 12391, 29161
Species Mouse, Rat
Purification Method purified
Accession Number P51637, P51638
Common Name Mouse/Rat Caveolin 3 (aa 1-18)
Concentration 0.5 mg/mL
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation peptide with no preservative
For Use With (Application) Control
Gene Symbol Cav3
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Sequence M(1) M T E E H T D L E A R I I K D I H(18) C
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.