missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse/Rat FGF-21 Quantikine ELISA Kit

Product Code. 18782334
Click to view available options
Quantity:
1 x 96-well Plate
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18782334

Brand: R&D Systems™ MF2100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse/Rat FGF-21 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse/Rat FGF-21 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse, rat FGF-21 in cell culture supernates (50 ul), serum (25 ul), edta plasma (25 ul), heparin plasma (25 ul).
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 31.30 - 2000pg/mL
Assay Sensitivity 13.4pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA), Plasma (Heparin), Serum
Specificity Natural and Recombinant Mouse and Rat FGF-21
For Use With (Application) ELISA
Gene ID (Entrez) 56636
Quantity 1 x 96-well Plate
Sample Volume Cell Culture Supernates (50uL),Serum (25uL),EDTA Plasma (25uL),Heparin Plasma (25uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym FGF21, FGF-21, fibroblast growth factor 21
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.