missing translation for 'onlineSavingsMsg'
Learn More

Mouse small intestine normal tissue lysate - total protein, 1 mg

Product Code. 32147901 Shop All Abcam Products
This item is not returnable. View return policy
Product Code. 32147901 Supplier Abcam Supplier No. ab297071mg
 Restricted Item
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse small intestine normal cell/tissue lysate.

  • Species: Mouse
  • Tissue: small intestine normal
  • Form: Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.