missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse SorCS1 (AAF68196.1) VersaClone cDNA

Product Code. 18702477
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18702477 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18702477 Supplier R&D Systems™ Supplier No. RDC3795
  • ONLINE EXCLUSIVE PRICE
    853.00 €
    806.40 € / 10µg
    Save 46.60 €
    5% Off
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Gene SorCS1
Accession Number AAF68196.1
Gene ID (Entrez) 114815
Gene Alias FLJ41758, FLJ43475, FLJ44957, hSorCS, SORCS, SORCS receptor 1, sorCS1, sortilin-related VPS10 domain containing receptor 1, VPS10 domain receptor protein SORCS 1, VPS10 domain receptor SorCS, VPS10 domain-containing receptor SorCS1
Content And Storage Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Product Line R&D Systems
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.