missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse TEM8/ANTXR1 (NP_473382) VersaClone cDNA
Description
- Also known as: Tem8; Antrx1; 2310008J16Rik; 2810405N18Rik
- ANTXR1 is a type I transmembrane protein and is a tumor-specific endothelial marker that has been implicated in colorectal cancer.
- It has been shown to also be a docking protein or receptor for Bacillus anthracis toxin, the causative agent of the disease, anthrax.
- The binding of the protective antigen (PA) component, of the tripartite anthrax toxin, to ANTXR1 mediates delivery of toxin components to the cytosol of cells.
- Once inside the cell, the other two components of anthrax toxin, edema factor (EF) and lethal factor (LF) disrupt normal cellular processes.
- The TEM8/ANTXR1 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mAntxr1sity |
| Accession Number | NP_473382 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1702 bp |
| Gene ID (Entrez) | 84168 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?