Learn More
ACROBiosystems Mouse Transferrin R2 Protein, mouse IgG1 Fc Tag, low endotoxin
Description
Transferrin receptor protein 2 (TFR2) is a single-pass type II membrane protein with a protease associated (PA) domain, an M28 peptidase domain and a transferrin receptor-like dimerization domain. The mutations in TFR2 gene have been associated with hereditary hemochromatosis type III. Furthermore, TFR2 is involved in the uptake of transferrin-bound iron into cells by endocytosis, although its role is minor compared to transferrin receptor 1.
Specifications
Specifications
| Accession Number | NP_056614.3 |
| Molecular Weight (g/mol) | 101.1 kDa |
| Quantity | 50 μg |
| Source | Mouse TFR2, Mouse Fc Tag (TF2-M5269) is expressed from human 293 cells (HEK293). It contains AA Arg 103 - Phe 798 (Accession # Q9JK x 3-1). |
| Immunogen | Arg 103 - Phe 798 |
| Storage Requirements | -20°C |
| Gene Alias | Transferrin R2,TFR2 |
| Common Name | Transferrin R2 |
| Conjugate | Unconjugated |
| Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.