missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Mouse VCAM-1/CD106 ELISA Kit

Product Code. 16891626 Shop All Proteintech Products
Click to view available options
Target Species:
Mouse
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 16891626

Brand: Proteintech™ KE1003896T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KE10038 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The VCAM-1 ELISA kit is to be used to detect and quantify protein levels of endogenous VCAM-1.

  • The assay recognizes mouse VCAM-1. An antibody specific for VCAM-1 has been pre-coated onto the microwells. The VCAM-1 protein in samples is captured by the coated antibody after incubation. Following extensive washing, another antibody of biotinylated specific for VCAM-1 is added to detect the captured VCAM-1 protein.
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent. Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.Vascular cell adhesion molecule 1 (VCAM-1), also known as CD106, is a transmembrane glycoprotein belonging to the immunoglobulin gene superfamily.
  • VCAM-1 is expressed by cytokine-activated endothelium, interacts with integrin VLA4 (α4β1) present on the surface of leukocytes, and mediates both adhesion and signal transduction.
  • The VCAM-1/integrin VLA4 (α4β1) interaction may play a pathophysiologic role both in immune responses and in leukocyte emigration to sites of inflammation.
  • VCAM-1 is also expressed either constitutively or inducibly in a variety of other cell types, including vascular smooth muscle cells, differentiating skeletal muscle cells, renal and neural epithelial cells, macrophages (Kupffer cells), dendritic cells, and bone marrow stromal cells.
  • Soluble VCAM-1 (sVCAM-1) has been found in addition to membrane-bound VCAM-1.

Specifications

Accession Number BC011159
Assay Range 15.6 to 1000 pg/mL
Assay Sensitivity 1.5 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Mouse
Gene ID (Entrez) 22329
Host Species Mouse
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID P29533
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Mouse VCAM-1/CD106 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.