missing translation for 'onlineSavingsMsg'
Learn More

Proteintech™ Mouse VEGF ELISA Kit

Product Code. 16821606 Shop All Proteintech Products
Click to view available options
Target Species:
Mouse
Unit Size:
96 tests
This item is not returnable. View return policy

Product Code. 16821606

Brand: Proteintech™ KE1000996T

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

KE10009 is a solid phase sandwich Enzyme Linked-Immuno-Sorbent Assay (Sandwich ELISA). The Mouse VEGF ELISA kit is to be used to detect and quantify protein levels of endogenous mouse VEGF.

  • Vascular endothelial growth factor (VEGF), is a signal protein produced by cells that stimulates vasculogenesis and angiogenesis.
  • It is part of the system that restores the oxygen supply to tissues when blood circulation is inadequate such as in hypoxic conditions. Serum concentration of VEGF is high in bronchial asthma and diabetes mellitus.
  • The activities of VEGF are not limited to the vascular system; VEGF plays a role in normal physiological functions such as bone formation, hematopoiesis, wound healing, and development.
  • Disruption of this gene in mice resulted in abnormal embryonic blood vessel formation.
  • VEGF is upregulated in many known tumors and its expression is correlated with tumor stage and progression.
  • The assay recognizes mouse VEGF. An antibody specific for mouse VEGF has been pre-coated onto the microwells. The mouse VEGF protein in samples is captured by the coated antibody after incubation.
  • Following extensive washing, another antibody of biotinylated specific for mouse VEGF is added to detect the captured mouse VEGF protein.
  • For signal development, Streptavidin-HRP is added, followed by Tetramethyl-benzidine (TMB) reagent.
  • Solution containing sulfuric acid is used to stop color development and the color intensity which is proportional to the quantity of bound protein is measurable at 450 nm with the correction wavelength set at 630 nm.

Specifications

Accession Number NP_001020421
Assay Range 7.8 to 500 pg/mL
Assay Sensitivity 2 pg/mL
Certifications/Compliance ISO 13485
Detection Method Sandwich ELISA, Double Antibody
Immunoassay Kit Format Coated ELISA Kit
Sample Type Plasma, Serum
Substrate DAB
Target Species Mouse
Gene ID (Entrez) 22339
Host Species Mouse
Immunoassay Kit Grade Solid
Interassay CV <10%
Intraassay CV <10%
Shipping Condition Blue ice
Storage Requirements -20°C to +4°C
Total Assay Time 4.5 hr.
UniProt ID Q00731
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 96TEST Human FOLR1 ELISA Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P234-Keep only in original container.
  • P264-Wash skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P362-Take off contaminated clothing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Sulphuric Acid, Tetramethylbenzidine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.