missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse VEGF Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit (1 Plate)
6 Plates
Unit Size:
1 set
Description
The Mouse VEGF Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies mouse VEGF in cell culture supernates (50 ul), tissue homogenates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul).
Specifications
Specifications
| Assay Range | 7.80 to 500 pg/mL |
| Assay Sensitivity | 3 pg/mL |
| Conjugate | HRP |
| Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Tissue Homogenates |
| Specificity | 164 and 120 amino acid residue forms of Mouse VEGF |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 22339 |
| Quantity | 1 Kit (1 Plate) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction