missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse VSTM2A (NP_001277468) VersaClone cDNA

Product Code. 18765828
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18765828 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18765828 Supplier R&D Systems™ Supplier No. RDC2283

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R&D Systems™ Mouse VSTM2A (NP_001277468) VersaClone cDNA plays a role in the regulation of the early stage of white and brown preadipocyte cell differentiation.

The Mouse VSTM2A (NP_001277468) VersaClone cDNA clone contains the complete ORF for VSTM2A, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The VSTM2A insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Specifications

Gene VSTM2A
Accession Number NP_001277468
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 772bp
pH 8.5
Gene ID (Entrez) 222008
Gene Alias MGC33530, V-set and transmembrane domain containing 2, V-set and transmembrane domain containing 2 A, V-set and transmembrane domain-containing protein 2 A, VSTM2
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.