missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Aldehyde Dehydrogenase, Potassium Activated

Product Code. 11446091 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 U
250 U
Unit Size:
100 units
250 units
Product Code. Quantity unitSize
11446091 100 U 100 units
11456091 250 U 250 units
2 options
This item is not returnable. View return policy

Product Code. 11446091

Brand: MP Biomedicals™ 0219067980

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

From yeast

Activity: 1U/mg solid (approx. 5U/mg protein). ADH Less than 0.1%. Unit Definition: One unit causes the oxidation of one micromole of acetaldehyde to acetic acid per minute at pH 8.0 and 25°C in the presence of potassium ions and thiols. Freeze dried stabilized powder.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≈5 U/mg Protein, ≥1 U/mg Solid
Color Off-White
pH 9
For Use With (Application) Oxidizes a number of Aliphatic and Aromatic Aldehydes
Content And Storage -20°C
Source Yeast
Quantity 100 U
Product Type Aldehyde Dehydrogenase
Molecular Weight (g/mol) ≈200000
Inhibitors SH-reagents, such as N-ethylmaleimide, 2-iodobenzoic Acid, Iodoacetic Acid and Chloromercuribenzoic Acid, Heavy Metals (especially Cu2-) and Acetaldehyde (at high concentrations)
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.