missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ alpha-Amylase from Bacillus subtilis

Product Code. 11463130 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463130 100 g 100g
11473130 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463130 Supplier MP Biomedicals™ Supplier No. 0210044780
  • ONLINE EXCLUSIVE PRICE
    119.97 €
    106.05 € / 100g
    Save 13.92 €
    11% Off
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Liquefying diastase supplied as dry, free-flowing powder with high uniformity and storage stability

Deactivation temperature: 90 to 100°C
TRUSTED_SUSTAINABILITY

Specifications

Color Off-White to Tan
Incubator Temperature 60°C to 80°C
pH 5 to 7
For Use With (Application) Carbohydrate hydrolysis
Content And Storage 4°C
Source Bacillus subtilis
Quantity 100 g
Product Type alpha-Amylase
Form Powder
compliance-icons
Product Identifier
  • amylase, alpha-
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: alpha-amylase
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.